Product Details
- SNP ID
-
rs77434419
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:54063808 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACTTAATCTCTAGAAGATCAGAT[A/C]TCTCACTTAACCCTCTCACTTCTGT
- Phenotype
-
MIM: 142974
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ12825
PubMed Links
Gene Details
- Gene
- FLJ12825
- Gene Name
- uncharacterized LOC440101
There are no transcripts associated with this gene.
- Gene
- HOXC4
- Gene Name
- homeobox C4
There are no transcripts associated with this gene.
View Full Product Details