Product Details

SNP ID
rs77427130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54499448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCAACAAGAAATTTCCCAACATA[C/G]ATGTCCGAAACAGCACGGTGAGAAC
Phenotype
MIM: 141180
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NCKAP1L PubMed Links

Gene Details

Gene
NCKAP1L
Gene Name
NCK associated protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184976.1 146 Missense Mutation CAT,GAT H16D NP_001171905.1
NM_005337.4 146 Missense Mutation CAT,GAT H66D NP_005328.2

View Full Product Details