Product Details

SNP ID
rs75790912
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55331962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGCAATTATCACCAGGAATAAG[A/T]CTATTTCCTATAACAACTGTGCAGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR6C3 PubMed Links

Gene Details

Gene
OR6C3
Gene Name
olfactory receptor family 6 subfamily C member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054104.1 262 Missense Mutation ACT,TCT T88S NP_473445.1

View Full Product Details