Product Details

SNP ID
rs74832951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57489985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACCTCCCTGGTGAGAACTGTGC[A/G]TATCACTCCAACCCTAGGAGCTTGG
Phenotype
MIM: 610576 MIM: 156560
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP9 PubMed Links

Gene Details

Gene
ARHGAP9
Gene Name
Rho GTPase activating protein 9
There are no transcripts associated with this gene.

Gene
MARS
Gene Name
methionyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004990.3 Intron NP_004981.2
XM_006719398.3 Intron XP_006719461.1

View Full Product Details