Product Details

SNP ID
rs75850843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:58872784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCAAAGCTTGAATAGGCATCTA[C/G]GTGAGGTCTCCTGAGAGCTTTTCCA
Phenotype
MIM: 608870
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LRIG3 PubMed Links

Gene Details

Gene
LRIG3
Gene Name
leucine rich repeats and immunoglobulin like domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136051.2 3451 Missense Mutation CTA,GTA L990V NP_001129523.1
NM_153377.4 3451 Missense Mutation CTA,GTA L1050V NP_700356.2
XM_011537893.2 3451 Missense Mutation CTA,GTA L972V XP_011536195.1
XM_017018790.1 3451 Intron XP_016874279.1

View Full Product Details