Product Details
- SNP ID
-
rs78573387
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:71613374 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAATTAGCTGGCATAACTGAACAA[A/G]ACCGTTCCAAGGTTTTGGAATGCTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZFC3H1
PubMed Links
Gene Details
- Gene
- ZFC3H1
- Gene Name
- zinc finger C3H1-type containing
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144982.4 |
5947 |
Missense Mutation |
TCT,TTT |
S1863F |
NP_659419.3 |
View Full Product Details