Product Details
- SNP ID
-
rs75478848
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:85283611 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTAAAGTAGAAGGACAGCCCCTTC[A/G]CACCGAACTGAATAGAGCTATGGAC
- Phenotype
-
MIM: 601527
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ALX1
PubMed Links
Gene Details
- Gene
- ALX1
- Gene Name
- ALX homeobox 1
View Full Product Details