Product Details

SNP ID
rs78918249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:88042826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTTTGTTATTACTTTTTTAGA[C/T]CAGCCATACCTTTGGGCTCGACTAG
Phenotype
MIM: 610142
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf29 PubMed Links

Gene Details

Gene
C12orf29
Gene Name
chromosome 12 open reading frame 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009894.2 Intron NP_001009894.2
Gene
CEP290
Gene Name
centrosomal protein 290
There are no transcripts associated with this gene.

View Full Product Details