Product Details

SNP ID
rs79943404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:93468498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGTGACTGACTCTAAAGTCCT[G/T]GGAAGCAAGACGTTTAAATTTTGTA
Phenotype
MIM: 611847
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MRPL42 PubMed Links

Gene Details

Gene
MRPL42
Gene Name
mitochondrial ribosomal protein L42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014050.3 Intron NP_054769.1
NM_172177.3 Intron NP_751917.1

View Full Product Details