Product Details

SNP ID
rs77046066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:95079409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGATTCAGCCAGGTCTTAGAAGA[C/T]AGAATAGCCTGGCTGATATTTTTTA
Phenotype
MIM: 613520 MIM: 601529
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGD6 PubMed Links

Gene Details

Gene
FGD6
Gene Name
FYVE, RhoGEF and PH domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018351.3 6628 UTR 3 NP_060821.3
Gene
NR2C1
Gene Name
nuclear receptor subfamily 2 group C member 1
There are no transcripts associated with this gene.

View Full Product Details