Product Details

SNP ID
rs75401321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:96278306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGAATTAAGCCTATGAAAAGCT[A/G]TAATGTCAAATTTGCAACAAATCAA
Phenotype
MIM: 603440 MIM: 600247
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDK17 PubMed Links

Gene Details

Gene
CDK17
Gene Name
cyclin dependent kinase 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170464.2 4140 UTR 3 NP_001163935.1
NM_002595.4 4140 UTR 3 NP_002586.2
XM_017019405.1 4140 Intron XP_016874894.1
XM_017019406.1 4140 Intron XP_016874895.1
XM_017019407.1 4140 Intron XP_016874896.1
Gene
ELK3
Gene Name
ELK3, ETS transcription factor
There are no transcripts associated with this gene.

View Full Product Details