Product Details

SNP ID
rs76789711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:100719367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAACCTAGTGAACAAGTTAAGGA[A/G]AAAAACATTGAGCACATTTAATACA
Phenotype
MIM: 610111
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANO4 PubMed Links

Gene Details

Gene
ANO4
Gene Name
anoctamin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286615.1 Intron NP_001273544.1
NM_001286616.1 Intron NP_001273545.1
NM_178826.3 Intron NP_849148.2
XM_011537911.2 Intron XP_011536213.1
XM_011537912.2 Intron XP_011536214.1
XM_011537913.2 Intron XP_011536215.1
XM_011537914.2 Intron XP_011536216.1
XM_011537915.2 Intron XP_011536217.1
XM_011537916.2 Intron XP_011536218.1
XM_011537918.2 Intron XP_011536220.1
XM_017018808.1 Intron XP_016874297.1
XM_017018809.1 Intron XP_016874298.1
XM_017018810.1 Intron XP_016874299.1
XM_017018811.1 Intron XP_016874300.1
XM_017018812.1 Intron XP_016874301.1
XM_017018813.1 Intron XP_016874302.1
XM_017018814.1 Intron XP_016874303.1
XM_017018815.1 Intron XP_016874304.1
XM_017018816.1 Intron XP_016874305.1
XM_017018817.1 Intron XP_016874306.1

View Full Product Details