Product Details

SNP ID
rs78240090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:101281124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATACTTTCCCTTCCAGTTTCTTAC[A/G]TTTGCTGAACGACTGGGGAATGTTA
Phenotype
MIM: 612822
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UTP20 PubMed Links

Gene Details

Gene
UTP20
Gene Name
UTP20, small subunit processome component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014503.2 210 Silent Mutation ACA,ACG T18T NP_055318.2

View Full Product Details