Product Details
- SNP ID
-
rs78828803
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:101395427 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTAACACAAGAAAGCAAGAAAAGA[A/G]TATTTTACATTACATAGAACATTCA
- Phenotype
-
MIM: 603425
MIM: 612822
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148098547] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARL1
- Gene Name
- ADP ribosylation factor like GTPase 1
- Gene
- UTP20
- Gene Name
- UTP20, small subunit processome component
There are no transcripts associated with this gene.
View Full Product Details