Product Details
- SNP ID
-
rs79850286
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:102077980 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATCAAATGTAAAGGAAGAGGCTA[C/T]GTAAAAATCTCTAGAGAATATAAAA
- Phenotype
-
MIM: 609264
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NUP37
PubMed Links
Gene Details
- Gene
- NUP37
- Gene Name
- nucleoporin 37
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024057.3 |
|
Intron |
|
|
NP_076962.2 |
View Full Product Details