Product Details

SNP ID
rs75343959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:107776524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTCTCAATTGTTTTGTCCACTG[G/T]TTTCTTCATTTTCCTTTTCTCTGCA
Phenotype
MIM: 609155
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ASCL4 PubMed Links

Gene Details

Gene
ASCL4
Gene Name
achaete-scute family bHLH transcription factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203436.2 2140 UTR 3 NP_982260.2

View Full Product Details