Product Details

SNP ID
rs74342954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:46358927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGTTCAGGAACCTCAAGGTAGA[C/T]AAGATAGCTCCCAGAAAATCATCCA
Phenotype
MIM: 605180
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC38A2 PubMed Links

Gene Details

Gene
SLC38A2
Gene Name
solute carrier family 38 member 2
There are no transcripts associated with this gene.

View Full Product Details