Product Details

SNP ID
rs78843541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196222151 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATTATGGGTGATTACTCTTTTT[C/T]CAAATTCTTATTTAATGTTCCTGTT
Phenotype
MIM: 612084
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC51A PubMed Links

Gene Details

Gene
SLC51A
Gene Name
solute carrier family 51 alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152672.5 Intron NP_689885.4

View Full Product Details