Product Details

SNP ID
rs78851703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:197753112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGTCCCTCTATTGCCTTTCTTT[A/T]GGTGAGAATTTTACAGAAATGGGAT
Phenotype
MIM: 616933 MIM: 613516
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FYTTD1 PubMed Links

Gene Details

Gene
FYTTD1
Gene Name
forty-two-three domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011537.2 Intron NP_001011537.2
NM_032288.6 Intron NP_115664.2
Gene
RUBCN
Gene Name
RUN and cysteine rich domain containing beclin 1 interacting protein
There are no transcripts associated with this gene.

View Full Product Details