Product Details

SNP ID
rs76471934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:59959349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAATGTTTATTGAAGAAAATGAA[G/T]GAATGAGTAGAAATAAAAGAATGAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC01553 PubMed Links

Gene Details

Gene
LINC01553
Gene Name
long intergenic non-protein coding RNA 1553
There are no transcripts associated with this gene.

View Full Product Details