Product Details

SNP ID
rs76270630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:63176970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTAATCAGTTAAATAATCTGCTT[C/T]AGAAGGCACAAATGATCATACTTCA
Phenotype
MIM: 604503
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JMJD1C PubMed Links

Gene Details

Gene
JMJD1C
Gene Name
jumonji domain containing 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282948.1 Intron NP_001269877.1
NM_001318153.1 Intron NP_001305082.1
NM_001318154.1 Intron NP_001305083.1
NM_001322252.1 Intron NP_001309181.1
NM_001322254.1 Intron NP_001309183.1
NM_001322258.1 Intron NP_001309187.1
NM_032776.2 Intron NP_116165.1
XM_011539503.1 Intron XP_011537805.1
XM_011539508.2 Intron XP_011537810.1
XM_017015897.1 Intron XP_016871386.1
XM_017015898.1 Intron XP_016871387.1
XM_017015899.1 Intron XP_016871388.1
XM_017015900.1 Intron XP_016871389.1
XM_017015901.1 Intron XP_016871390.1
XM_017015902.1 Intron XP_016871391.1
XM_017015903.1 Intron XP_016871392.1

View Full Product Details