Product Details

SNP ID
rs76342145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:68562742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGAGGGTTGGGTCTCCACTGTC[C/T]CTGCCCTTTGTTTCTCTTCCTTCTC
Phenotype
MIM: 607790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TET1 PubMed Links

Gene Details

Gene
TET1
Gene Name
tet methylcytosine dioxygenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030625.2 Intron NP_085128.2
XM_011540204.2 Intron XP_011538506.1
XM_011540205.2 Intron XP_011538507.1
XM_011540206.2 Intron XP_011538508.1
XM_011540207.2 Intron XP_011538509.1
XM_017016686.1 Intron XP_016872175.1
XM_017016687.1 Intron XP_016872176.1
XM_017016688.1 Intron XP_016872177.1
XM_017016689.1 Intron XP_016872178.1

View Full Product Details