Product Details

SNP ID
rs75168453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70066560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTGAGAATAACCCCTCTGCAAA[C/T]GGTTCTCTGGGAAAAACAGGGTCTC
Phenotype
MIM: 616141
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
H2AFY2 PubMed Links

Gene Details

Gene
H2AFY2
Gene Name
H2A histone family member Y2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018649.2 Intron NP_061119.1

View Full Product Details