Product Details

SNP ID
rs74720353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:71748499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTCAATTCCATTCCTTCAGCCT[C/T]TCCCTGGGAGGGAATAAAGGCAGGA
Phenotype
MIM: 615608 MIM: 605516
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf54 PubMed Links

Gene Details

Gene
C10orf54
Gene Name
chromosome 10 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022153.1 3831 UTR 3 NP_071436.1
Gene
CDH23
Gene Name
cadherin-related 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171930.1 3831 Intron NP_001165401.1
NM_001171931.1 3831 Intron NP_001165402.1
NM_001171932.1 3831 Intron NP_001165403.1
NM_001171933.1 3831 Intron NP_001165404.1
NM_001171934.1 3831 Intron NP_001165405.1
NM_001171935.1 3831 Intron NP_001165406.1
NM_001171936.1 3831 Intron NP_001165407.1
NM_022124.5 3831 Intron NP_071407.4
NM_052836.3 3831 Intron NP_443068.1
XM_006717940.3 3831 Intron XP_006718003.1
XM_006717942.3 3831 Intron XP_006718005.1
XM_011540039.2 3831 Intron XP_011538341.1
XM_011540042.2 3831 Intron XP_011538344.1
XM_011540043.2 3831 Intron XP_011538345.1
XM_011540044.2 3831 Intron XP_011538346.1
XM_011540045.2 3831 Intron XP_011538347.1
XM_011540046.2 3831 Intron XP_011538348.1
XM_011540047.2 3831 Intron XP_011538349.1
XM_011540048.2 3831 Intron XP_011538350.1
XM_011540049.2 3831 Intron XP_011538351.1
XM_011540051.2 3831 Intron XP_011538353.1
XM_011540052.2 3831 Intron XP_011538354.1
XM_017016499.1 3831 Intron XP_016871988.1
XM_017016500.1 3831 Intron XP_016871989.1
XM_017016501.1 3831 Intron XP_016871990.1
XM_017016502.1 3831 Intron XP_016871991.1
XM_017016503.1 3831 Intron XP_016871992.1
XM_017016504.1 3831 Intron XP_016871993.1
XM_017016505.1 3831 Intron XP_016871994.1
XM_017016506.1 3831 Intron XP_016871995.1
XM_017016507.1 3831 Intron XP_016871996.1
XM_017016508.1 3831 Intron XP_016871997.1

View Full Product Details