Product Details

SNP ID
rs76512705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:72718745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGAATTTTCCAAGTGCCCATCA[A/G]CAGTAGAATAAGTATATAATTTGTG
Phenotype
MIM: 614197
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MCU PubMed Links

Gene Details

Gene
MCU
Gene Name
mitochondrial calcium uniporter
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270679.1 Intron NP_001257608.1
NM_001270680.1 Intron NP_001257609.1
NM_138357.2 Intron NP_612366.1
XM_017016882.1 Intron XP_016872371.1
Gene
MIR4676
Gene Name
microRNA 4676
There are no transcripts associated with this gene.

View Full Product Details