Product Details

SNP ID
rs79860187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:76870851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGAATATGGACAAATAAGCAGA[A/G]TTCCTGCCCTTCGAGTGAGTCTTTT
Phenotype
MIM: 600150
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNMA1 PubMed Links

Gene Details

Gene
KCNMA1
Gene Name
potassium calcium-activated channel subfamily M alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014797.2 10114 UTR 3 NP_001014797.1
NM_001161352.1 10114 Intron NP_001154824.1
NM_001161353.1 10114 Intron NP_001154825.1
NM_001271518.1 10114 UTR 3 NP_001258447.1
NM_001271519.1 10114 Intron NP_001258448.1
NM_001271520.1 10114 Intron NP_001258449.1
NM_001271521.1 10114 Intron NP_001258450.1
NM_001271522.1 10114 Intron NP_001258451.1
NM_001322829.1 10114 UTR 3 NP_001309758.1
NM_001322830.1 10114 UTR 3 NP_001309759.1
NM_001322832.1 10114 UTR 3 NP_001309761.1
NM_001322835.1 10114 UTR 3 NP_001309764.1
NM_001322836.1 10114 Intron NP_001309765.1
NM_001322837.1 10114 Intron NP_001309766.1
NM_001322838.1 10114 UTR 3 NP_001309767.1
NM_001322839.1 10114 Intron NP_001309768.1
NM_002247.3 10114 Intron NP_002238.2
XM_005269776.3 10114 UTR 3 XP_005269833.1
XM_005269778.1 10114 UTR 3 XP_005269835.1
XM_005269781.2 10114 UTR 3 XP_005269838.1
XM_005269787.3 10114 UTR 3 XP_005269844.1
XM_005269789.2 10114 UTR 3 XP_005269846.1
XM_005269792.2 10114 UTR 3 XP_005269849.1
XM_005269796.2 10114 UTR 3 XP_005269853.1
XM_006717826.2 10114 Intron XP_006717889.1
XM_011539773.1 10114 UTR 3 XP_011538075.1
XM_011539774.1 10114 Intron XP_011538076.1
XM_011539775.1 10114 UTR 3 XP_011538077.1
XM_011539776.1 10114 UTR 3 XP_011538078.1
XM_011539777.1 10114 Intron XP_011538079.1
XM_011539778.2 10114 Intron XP_011538080.1
XM_011539779.1 10114 Intron XP_011538081.1
XM_011539780.1 10114 UTR 3 XP_011538082.1
XM_011539781.2 10114 UTR 3 XP_011538083.1
XM_011539782.2 10114 Intron XP_011538084.1
XM_011539783.1 10114 Intron XP_011538085.1
XM_011539784.1 10114 Intron XP_011538086.1
XM_011539785.1 10114 UTR 3 XP_011538087.1
XM_017016207.1 10114 UTR 3 XP_016871696.1
XM_017016208.1 10114 UTR 3 XP_016871697.1
XM_017016209.1 10114 Intron XP_016871698.1
XM_017016210.1 10114 UTR 3 XP_016871699.1
XM_017016211.1 10114 UTR 3 XP_016871700.1
XM_017016212.1 10114 UTR 3 XP_016871701.1
XM_017016213.1 10114 UTR 3 XP_016871702.1
XM_017016214.1 10114 Intron XP_016871703.1
XM_017016215.1 10114 Intron XP_016871704.1
XM_017016216.1 10114 UTR 3 XP_016871705.1
XM_017016217.1 10114 UTR 3 XP_016871706.1
XM_017016218.1 10114 Intron XP_016871707.1
XM_017016219.1 10114 Intron XP_016871708.1
XM_017016220.1 10114 Intron XP_016871709.1
XM_017016221.1 10114 Intron XP_016871710.1
XM_017016222.1 10114 Intron XP_016871711.1
XM_017016223.1 10114 UTR 3 XP_016871712.1

View Full Product Details