Product Details

SNP ID
rs74332561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:88820758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGATAGCCAGAAAATATCTAGAC[A/G]TTCTCTATATCATTCAGGTAAATCT
Phenotype
MIM: 613923
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD22 PubMed Links

Gene Details

Gene
ANKRD22
Gene Name
ankyrin repeat domain 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144590.2 3127 UTR 3 NP_653191.2
Gene
LIPM
Gene Name
lipase family member M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128215.1 3127 Intron NP_001121687.1
XM_011539748.2 3127 UTR 3 XP_011538050.1
XM_011539751.2 3127 UTR 3 XP_011538053.1
XM_011539752.2 3127 UTR 3 XP_011538054.1
XM_011539753.2 3127 Intron XP_011538055.1

View Full Product Details