Product Details

SNP ID
rs77786357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:93567760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGGCTCCTTCCAGAAGTTTACC[C/T]GTAATTCAAGGAGGAGTTGGGCTCC
Phenotype
MIM: 609044
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FFAR4 PubMed Links

Gene Details

Gene
FFAR4
Gene Name
free fatty acid receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195755.1 Intron NP_001182684.1
NM_181745.3 Intron NP_859529.2
XM_011539746.2 Intron XP_011538048.1

View Full Product Details