Product Details

SNP ID
rs75584607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:98457852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCCTCTCTCTCCAGTTGCTTCC[A/C]TGCTGAGCACAGCCACTGACCTGGT
Phenotype
MIM: 613469
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HPSE2 PubMed Links

Gene Details

Gene
HPSE2
Gene Name
heparanase 2 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166244.1 3427 UTR 3 NP_001159716.1
NM_001166245.1 3427 UTR 3 NP_001159717.1
NM_001166246.1 3427 UTR 3 NP_001159718.1
NM_021828.4 3427 UTR 3 NP_068600.4
XM_006717937.2 3427 UTR 3 XP_006718000.1
XM_011540029.1 3427 UTR 3 XP_011538331.1
XM_011540030.1 3427 UTR 3 XP_011538332.1
XM_011540031.2 3427 UTR 3 XP_011538333.1
XM_011540033.2 3427 UTR 3 XP_011538335.1
XM_017016495.1 3427 Intron XP_016871984.1
XM_017016496.1 3427 Intron XP_016871985.1
XM_017016497.1 3427 UTR 3 XP_016871986.1
XM_017016498.1 3427 UTR 3 XP_016871987.1
Gene
LOC101927278
Gene Name
uncharacterized LOC101927278
There are no transcripts associated with this gene.

View Full Product Details