Product Details

SNP ID
rs79218728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102655227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGAACGCAGCGCAGGCGCTGCAC[C/T]TCGGGGAGCGCATGCAGGAGGCCAA
Phenotype
MIM: 606125
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM8 PubMed Links

Gene Details

Gene
TRIM8
Gene Name
tripartite motif containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030912.2 937 Missense Mutation CTC,TTC L272F NP_112174.2

View Full Product Details