Product Details

SNP ID
rs78748561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103374631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAAAATGGATATTTGGGGAGAG[C/T]GAAGCCTTTTTGCTACAGAGTCATT
Phenotype
MIM: 601787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAF5 PubMed Links

Gene Details

Gene
TAF5
Gene Name
TATA-box binding protein associated factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006951.4 Intron NP_008882.2
NM_139052.2 Intron NP_620640.1

View Full Product Details