Product Details

SNP ID
rs76451872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:104272449 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTGATAAAGACTAAGATACTGG[C/T]TGAGATTGTTGCCGCTGGAGTCAAA
Phenotype
MIM: 605482 MIM: 612314
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTO1 PubMed Links

Gene Details

Gene
GSTO1
Gene Name
glutathione S-transferase omega 1
There are no transcripts associated with this gene.

Gene
GSTO2
Gene Name
glutathione S-transferase omega 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191013.1 Intron NP_001177942.1
NM_001191014.1 Intron NP_001177943.1
NM_001191015.1 Intron NP_001177944.1
NM_183239.1 Intron NP_899062.1
XM_006717624.3 Intron XP_006717687.1
XM_011539270.2 Intron XP_011537572.1
XM_011539271.1 Intron XP_011537573.1
XM_011539272.2 Intron XP_011537574.1
XM_011539273.2 Intron XP_011537575.1
XM_017015670.1 Intron XP_016871159.1
XM_017015671.1 Intron XP_016871160.1
Gene
MIR4482
Gene Name
microRNA 4482
There are no transcripts associated with this gene.

View Full Product Details