Product Details

SNP ID
rs74459175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:53802932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAAGCAGTGAAGGTGTTGATAGA[C/T]TTTTCAAACTATAGACAATGTTCGA
Phenotype
MIM: 605514
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDH15 PubMed Links

Gene Details

Gene
PCDH15
Gene Name
protocadherin related 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142763.1 9091 Intron NP_001136235.1
NM_001142764.1 9091 Intron NP_001136236.1
NM_001142765.1 9091 Intron NP_001136237.1
NM_001142766.1 9091 Intron NP_001136238.1
NM_001142767.1 9091 Intron NP_001136239.1
NM_001142768.1 9091 Intron NP_001136240.1
NM_001142769.1 9091 Intron NP_001136241.1
NM_001142770.1 9091 Intron NP_001136242.1
NM_001142771.1 9091 UTR 3 NP_001136243.1
NM_001142772.1 9091 UTR 3 NP_001136244.1
NM_001142773.1 9091 Intron NP_001136245.1
NM_033056.3 9091 Intron NP_149045.3
XM_017016571.1 9091 Intron XP_016872060.1
XM_017016572.1 9091 UTR 3 XP_016872061.1
XM_017016573.1 9091 Intron XP_016872062.1

View Full Product Details