Product Details

SNP ID
rs78476354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:112309140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTCCATGGAGATGCATCCATCCC[A/G]CTAAGTTCCTCGATGTCTCTATCAG
Phenotype
MIM: 602653
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GUCY2GP PubMed Links

Gene Details

Gene
GUCY2GP
Gene Name
guanylate cyclase 2G, pseudogene
There are no transcripts associated with this gene.

Gene
MIR6715A
Gene Name
microRNA 6715a
There are no transcripts associated with this gene.

Gene
MIR6715B
Gene Name
microRNA 6715b
There are no transcripts associated with this gene.

Gene
TECTB
Gene Name
tectorin beta
There are no transcripts associated with this gene.

View Full Product Details