Product Details

SNP ID
rs76420054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119032104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAATACACAAATGCATTACTAT[A/C]TGAAGATACCAGCAAGAGTTTAGTC
Phenotype
MIM: 602039 MIM: 608226
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EIF3A PubMed Links

Gene Details

Gene
EIF3A
Gene Name
eukaryotic translation initiation factor 3 subunit A
There are no transcripts associated with this gene.

Gene
NANOS1
Gene Name
nanos C2HC-type zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199461.3 2389 UTR 3 NP_955631.1

View Full Product Details