Product Details

SNP ID
rs75613220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122994009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGGGTGCTTGGCTGGCTGTTT[C/T]CTATGCTGGGAGTGCTCGTGTGGGC
Phenotype
MIM: 606238 MIM: 611310
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IKZF5 PubMed Links

Gene Details

Gene
IKZF5
Gene Name
IKAROS family zinc finger 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271840.1 1830 Missense Mutation GAA,GGA E344G NP_001258769.1
XM_006717947.3 1830 Missense Mutation GAA,GGA E344G XP_006718010.1
XM_017016550.1 1830 Missense Mutation GAA,GGA E344G XP_016872039.1
XM_017016551.1 1830 Missense Mutation GAA,GGA E344G XP_016872040.1
XM_017016552.1 1830 Missense Mutation GAA,GGA E344G XP_016872041.1
XM_017016553.1 1830 Missense Mutation GAA,GGA E276G XP_016872042.1
XM_017016554.1 1830 Missense Mutation GAA,GGA E276G XP_016872043.1
XM_017016555.1 1830 Missense Mutation GAA,GGA E276G XP_016872044.1
Gene
PSTK
Gene Name
phosphoseryl-tRNA kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153336.2 1830 Intron NP_699167.2
XM_017015640.1 1830 Intron XP_016871129.1
XM_017015641.1 1830 Intron XP_016871130.1

View Full Product Details