Product Details

SNP ID
rs79549040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:123746382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTGCAATTTTAGCTGTGCAAAT[G/T]GCAAATGCACGTGGAACCCTTGCTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CPXM2 PubMed Links

Gene Details

Gene
CPXM2
Gene Name
carboxypeptidase X (M14 family), member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198148.2 2579 UTR 3 NP_937791.2
XM_005269528.3 2579 Intron XP_005269585.1
XM_011539283.2 2579 Intron XP_011537585.1
XM_011539285.2 2579 UTR 3 XP_011537587.1
XM_011539286.1 2579 UTR 3 XP_011537588.1
XM_017015673.1 2579 UTR 3 XP_016871162.1
XM_017015674.1 2579 UTR 3 XP_016871163.1

View Full Product Details