Product Details

SNP ID
rs77830073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:124759716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACATACCTCAAGAAGACATTGA[A/G]AGGACTCAATTACTTAATAAGACAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
METTL10 PubMed Links

Gene Details

Gene
METTL10
Gene Name
methyltransferase like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304467.1 1775 UTR 3 NP_001291396.1
NM_001304468.1 1775 UTR 3 NP_001291397.1
NM_212554.3 1775 UTR 3 NP_997719.2

View Full Product Details