Product Details

SNP ID
rs79266966
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:124765438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTTCCTTGGTCCAATTACATGA[C/T]GTTATTAGAAAAAAGCCTTTTACTT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
METTL10 PubMed Links

Gene Details

Gene
METTL10
Gene Name
methyltransferase like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304467.1 682 Silent Mutation ACA,ACG T112T NP_001291396.1
NM_001304468.1 682 Silent Mutation ACA,ACG T112T NP_001291397.1
NM_212554.3 682 Silent Mutation ACA,ACG T190T NP_997719.2

View Full Product Details