Product Details

SNP ID
rs77774010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:125576858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTCTGGTTCTCCCTGTGGGTCC[A/G]TTGCTGTCATTCTTTCCTGTGAGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEX36 PubMed Links

Gene Details

Gene
TEX36
Gene Name
testis expressed 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128202.2 436 Intron NP_001121674.1
NM_001318133.1 436 Missense Mutation ACG,ATG T94M NP_001305062.1
XM_005269817.4 436 Intron XP_005269874.1
Gene
TEX36-AS1
Gene Name
TEX36 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details