Product Details

SNP ID
rs79702543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:55599979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATGAAGAGGCGGGTATAAATAA[A/G]TAGGAGAAGACCCTGTGGAGCTTGA
Phenotype
MIM: 605514
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTRNR2L5 PubMed Links

Gene Details

Gene
MTRNR2L5
Gene Name
MT-RNR2-like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190478.1 990 UTR 3 NP_001177407.1
Gene
PCDH15
Gene Name
protocadherin related 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142763.1 990 Intron NP_001136235.1
NM_001142764.1 990 Intron NP_001136236.1
NM_001142765.1 990 Intron NP_001136237.1
NM_001142766.1 990 Intron NP_001136238.1
NM_001142767.1 990 Intron NP_001136239.1
NM_001142768.1 990 Intron NP_001136240.1
NM_001142769.1 990 Intron NP_001136241.1
NM_001142770.1 990 Intron NP_001136242.1
NM_001142771.1 990 Intron NP_001136243.1
NM_001142772.1 990 Intron NP_001136244.1
NM_001142773.1 990 Intron NP_001136245.1
NM_033056.3 990 Intron NP_149045.3
XM_017016571.1 990 Intron XP_016872060.1
XM_017016572.1 990 Intron XP_016872061.1
XM_017016573.1 990 Intron XP_016872062.1

View Full Product Details