Product Details
- SNP ID
-
hCV104718241
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:47367320 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTACAGACATCATCACAAACCTTG[T/G]TGACTCAGTGCTCAGGGTCTGCAGG
- Phenotype
-
MIM: 180290
- Polymorphism
- T/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RBP3
PubMed Links
Gene Details
- Gene
- RBP3
- Gene Name
- retinol binding protein 3
There are no transcripts associated with this gene.
- Gene
- ZNF488
- Gene Name
- zinc finger protein 488
There are no transcripts associated with this gene.
View Full Product Details