Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001105530.1 | 2031 | Intron | NP_001099000.1 | ||
NM_006367.3 | 2031 | Intron | NP_006358.1 | ||
XM_005270368.1 | 2031 | Intron | XP_005270425.1 | ||
XM_011540509.1 | 2031 | Intron | XP_011538811.1 | ||
XM_011540510.1 | 2031 | Intron | XP_011538812.1 | ||
XM_011540511.1 | 2031 | Intron | XP_011538813.1 | ||
XM_011540512.1 | 2031 | Intron | XP_011538814.1 | ||
XM_011540513.2 | 2031 | Intron | XP_011538815.1 | ||
XM_011540514.1 | 2031 | Intron | XP_011538816.1 | ||
XM_011540515.1 | 2031 | Intron | XP_011538817.1 | ||
XM_017000067.1 | 2031 | Intron | XP_016855556.1 | ||
XM_017000068.1 | 2031 | Intron | XP_016855557.1 | ||
XM_017000069.1 | 2031 | Intron | XP_016855558.1 | ||
XM_017000070.1 | 2031 | Intron | XP_016855559.1 | ||
XM_017000071.1 | 2031 | Intron | XP_016855560.1 | ||
XM_017000072.1 | 2031 | Intron | XP_016855561.1 | ||
XM_017000073.1 | 2031 | Intron | XP_016855562.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000310.3 | 2031 | UTR 3 | NP_000301.1 | ||
NM_001142604.1 | 2031 | UTR 3 | NP_001136076.1 | ||
XM_005271008.1 | 2031 | UTR 3 | XP_005271065.1 |