Product Details

SNP ID
rs76250039
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:40073605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATGCAGAAATAGCCAAGCAAGA[C/T]TTTTCCAAGCACTGGTTAAATCACC
Phenotype
MIM: 600722
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAP1 PubMed Links

Gene Details

Gene
CAP1
Gene Name
adenylate cyclase associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105530.1 1609 Intron NP_001099000.1
NM_006367.3 1609 Intron NP_006358.1
XM_005270368.1 1609 Intron XP_005270425.1
XM_011540509.1 1609 Intron XP_011538811.1
XM_011540510.1 1609 Intron XP_011538812.1
XM_011540511.1 1609 Intron XP_011538813.1
XM_011540512.1 1609 Intron XP_011538814.1
XM_011540513.2 1609 Intron XP_011538815.1
XM_011540514.1 1609 Intron XP_011538816.1
XM_011540515.1 1609 Intron XP_011538817.1
XM_017000067.1 1609 Intron XP_016855556.1
XM_017000068.1 1609 Intron XP_016855557.1
XM_017000069.1 1609 Intron XP_016855558.1
XM_017000070.1 1609 Intron XP_016855559.1
XM_017000071.1 1609 Intron XP_016855560.1
XM_017000072.1 1609 Intron XP_016855561.1
XM_017000073.1 1609 Intron XP_016855562.1
Gene
PPT1
Gene Name
palmitoyl-protein thioesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000310.3 1609 UTR 3 NP_000301.1
NM_001142604.1 1609 UTR 3 NP_001136076.1
XM_005271008.1 1609 UTR 3 XP_005271065.1

View Full Product Details