Product Details
- SNP ID
-
rs80008523
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:30712581 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCAAGAGGAGGCACAAACCCCTG[A/G]TACGACCTAAACCCACTCTCTTTTC
- Phenotype
-
MIM: 115437
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MATN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs151272601] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MATN1
- Gene Name
- matrilin 1, cartilage matrix protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002379.3 |
2526 |
UTR 3 |
|
|
NP_002370.1 |
- Gene
- MATN1-AS1
- Gene Name
- MATN1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details