Product Details

SNP ID
rs79526021
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:42458635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATAAATAAACTGGTATATGATCC[C/T]TATCACACTAACAGCTTTTTTTTAG
Phenotype
MIM: 609853
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPCS PubMed Links

Gene Details

Gene
PPCS
Gene Name
phosphopantothenoylcysteine synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077447.2 Intron NP_001070915.1
NM_001287506.1 Intron NP_001274435.1
NM_001287507.1 Intron NP_001274436.1
NM_001287508.1 Intron NP_001274437.1
NM_001287509.1 Intron NP_001274438.1
NM_001287510.1 Intron NP_001274439.1
NM_001287511.1 Intron NP_001274440.1
NM_024664.3 Intron NP_078940.2
Gene
ZMYND12
Gene Name
zinc finger MYND-type containing 12
There are no transcripts associated with this gene.

View Full Product Details