Product Details
- SNP ID
-
rs78541695
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
10
- Location
-
Chr.1:46609269 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACACACACACCCCGGCATCTGTGCT[C/T]ACTCACAGGCAGACTGCTCACTTTC
- Phenotype
-
MIM: 606724
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MKNK1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs543977500] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MKNK1
- Gene Name
- MAP kinase interacting serine/threonine kinase 1
There are no transcripts associated with this gene.
- Gene
- MOB3C
- Gene Name
- MOB kinase activator 3C
View Full Product Details