Product Details

SNP ID
rs76178225
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:47185517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGCATTCAACAACAACCTTCTC[A/G]TCTCTTACTCAAGTTTAAACCCCCT
Phenotype
MIM: 607178
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00853 PubMed Links

Gene Details

Gene
LINC00853
Gene Name
long intergenic non-protein coding RNA 853
There are no transcripts associated with this gene.

Gene
PDZK1IP1
Gene Name
PDZK1 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005764.3 Intron NP_005755.1

View Full Product Details