Product Details
- SNP ID
-
rs78198038
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
14
- Location
-
Chr.1:51790271 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACTCTTTTTCTCCACAAAATCTTG[C/T]AGACCTCAATATATAGCATAAAAAT
- Phenotype
-
MIM: 602651
MIM: 606737
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NRDC
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113912828] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NRDC
- Gene Name
- nardilysin convertase
- Gene
- OSBPL9
- Gene Name
- oxysterol binding protein like 9
There are no transcripts associated with this gene.
View Full Product Details