Product Details

SNP ID
rs74224608
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:32130314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGACTGCTTCACAGCTAGTAGA[A/G]AGGACACAAATGTGACTTTCTTGGC
Phenotype
MIM: 610563
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KPNA6 PubMed Links

Gene Details

Gene
KPNA6
Gene Name
karyopherin subunit alpha 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012316.4 Intron NP_036448.1
XM_005270711.3 Intron XP_005270768.1

View Full Product Details